Search
Search

DMTF1| Gene Symbol: DMTF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713840.3 | XP_002713886.1 | 8 - 9 | 934 | 84 | |
| XM_008261799.2 | XP_008260021.1 | 7 - 8 | 811 | 84 | ||
| XM_017344331.1 | XP_017199820.1 | 9 - 10 | 1067 | 84 | ||
| XM_017344332.1 | XP_017199821.1 | 8 - 9 | 935 | 84 | ||
Set Membership: |
|