Search
Search

SOX5| Gene Symbol: SOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259527.2 | XP_008257749.1 | 9 - 10 | 1301 | 86 | |
| XM_008259529.2 | XP_008257751.1 | 13 - 14 | 1693 | 86 | ||
| XM_008259534.2 | XP_008257756.1 | 9 - 10 | 1222 | 86 | ||
| XM_008259535.2 | XP_008257757.1 | 9 - 10 | 1718 | 86 | ||
| XM_008259536.2 | XP_008257758.1 | 5 - 6 | 537 | 86 | ||
| XM_008259537.2 | XP_008257759.1 | 5 - 6 | 534 | 86 | ||
| XM_008259538.2 | XP_008257760.1 | 5 - 6 | 551 | 86 | ||
| XM_008259539.2 | XP_008257761.1 | 5 - 6 | 548 | 86 | ||
| XM_017343332.1 | XP_017198821.1 | 9 - 10 | 1224 | 86 | ||
| XM_017343333.1 | XP_017198822.1 | 9 - 10 | 1221 | 86 | ||
| XM_017343334.1 | XP_017198823.1 | 9 - 10 | 1155 | 86 | ||
| XM_017343335.1 | XP_017198824.1 | 9 - 10 | 1119 | 86 | ||
| XM_017343336.1 | XP_017198825.1 | 9 - 10 | 1115 | 86 | ||
| XM_017343337.1 | XP_017198826.1 | 4 - 5 | 453 | 86 | ||
Set Membership: |
|