Search
Search

STK17A| Gene Symbol: STK17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082006.1 | NP_001075475.1 | 3 - 4 | 637 | 155 | |
| XM_008261482.2 | XP_008259704.1 | 4 - 5 | 802 | 155 | ||
| GenBank mRNA | AB042195.1 | - | 3 - 4 | 637 | 155 | |
Set Membership: |
|