Search
Search

SLC22A1| Gene Symbol: SLC22A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082022.1 | NP_001075491.1 | 10 - 11 | 1659 | 63 | |
| GenBank mRNA | AF015958.1 | - | 10 - 11 | 1659 | 63 | |
Set Membership: |
|