Search
Search

ITGB3| Gene Symbol: ITGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082066.1 | NP_001075535.1 | 14 - 15 | 2302 | 58 | |
| XM_008271093.2 | XP_008269315.1 | 14 - 15 | 3089 | 58 | ||
| GenBank mRNA | AF170529.1 | - | 14 - 15 | 2302 | 58 | |
Set Membership: |
|