Search
Search

VLDLR| Gene Symbol: VLDLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082188.1 | NP_001075657.1 | 11 - 12 | 1719 | 64 | |
| XM_008255864.2 | XP_008254086.1 | 12 - 13 | 1836 | 64 | ||
| XM_008255867.2 | XP_008254089.1 | 11 - 12 | 1802 | 64 | ||
| XM_008255870.2 | XP_008254092.1 | 10 - 11 | 1679 | 64 | ||
| XM_017347307.1 | XP_017202796.1 | 10 - 11 | 1679 | 64 | ||
| GenBank mRNA | D11100.1 | - | 11 - 12 | 1719 | 64 | |
Set Membership: |
|