Search
Search

CSNK1A1| Gene Symbol: CSNK1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082373.1 | NP_001075842.1 | 6 - 7 | 1056 | 78 | |
| XM_008254851.2 | XP_008253073.1 | 7 - 8 | 1351 | 78 | ||
| XM_008254854.2 | XP_008253076.1 | 7 - 8 | 1350 | 78 | ||
| XM_008254855.2 | XP_008253077.1 | 7 - 8 | 1344 | 78 | ||
| XM_008254856.2 | XP_008253078.1 | 7 - 8 | 1347 | 78 | ||
| XM_008254857.2 | XP_008253079.1 | 7 - 8 | 1351 | 78 | ||
| XM_008254858.2 | XP_008253080.1 | 7 - 8 | 1345 | 78 | ||
| XM_008254859.2 | XP_008253081.1 | 6 - 7 | 1250 | 78 | ||
| XM_008254860.2 | XP_008253082.1 | 6 - 7 | 1254 | 78 | ||
| GenBank mRNA | U59166.1 | - | 6 - 7 | 1056 | 78 | |
Set Membership: |
|