Search
Search

RYR2| Gene Symbol: RYR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082757.1 | NP_001076226.1 | 45 - 46 | 7427 | 79 | |
| GenBank mRNA | M59743.1 | - | 45 - 46 | 7417 | 79 | |
| U50465.1 | - | 45 - 46 | 7427 | 79 | ||
Set Membership: |
|