Search
Search

CTNNA1| Gene Symbol: CTNNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171142.1 | NP_001164613.1 | 13 - 14 | 2014 | 60 | |
| XM_008254892.2 | XP_008253114.1 | 13 - 14 | 2023 | 60 | ||
Set Membership: |
|