Search
Search

CYP2C| Gene Symbol: CYP2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171265.1 | NP_001164736.1 | 4 - 5 | 671 | 89 | |
| GenBank mRNA | D26152.1 | - | 4 - 5 | 671 | 89 | |
| K01523.1 | - | 3 - 4 | 384 | 89 | ||
Set Membership: |
|