Search
Search

MAP1A| Gene Symbol: MAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171311.1 | NP_001164782.1 | 2 - 3 | 8010 | 91 | |
| XM_008268819.2 | XP_008267041.1 | 5 - 6 | 8904 | 91 | ||
Set Membership: |
|