Search
Search

PHF20| Gene Symbol: PHF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171336.1 | NP_001164807.1 | 11 - 12 | 1829 | 67 | |
| XM_008255971.2 | XP_008254193.1 | 12 - 13 | 1970 | 67 | ||
| XM_017341622.1 | XP_017197111.1 | 11 - 12 | 1855 | 67 | ||
Set Membership: |
|