Search
Search

KIFBP| Gene Symbol: KIFBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718448.3 | XP_002718494.1 | 6 - 7 | 1056 | 93 | |
| XM_008270032.2 | XP_008268254.1 | 6 - 7 | 1068 | 93 | ||
Set Membership: |
|