Search
Search

ZBTB45| Gene Symbol: ZBTB45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721898.3 | XP_002721944.1 | 1428 | 68 | ||
| XM_008249031.2 | XP_008247253.1 | 1348 | 68 | |||
| XM_017337896.1 | XP_017193385.1 | 1640 | 68 | |||
Set Membership: |
|