Search
Search

KHDC4| Gene Symbol: KHDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715407.3 | XP_002715453.1 | 3 - 4 | 449 | 60 | |
| XM_008264331.2 | XP_008262553.1 | 3 - 4 | 449 | 60 | ||
| XM_017345814.1 | XP_017201303.1 | 2 - 3 | 377 | 60 | ||
| XM_017345815.1 | XP_017201304.1 | 3 - 4 | 449 | 60 | ||
| XM_017345816.1 | XP_017201305.1 | 3 - 4 | 449 | 60 | ||
Set Membership: |
|