Search
Search

FAM184A| Gene Symbol: FAM184A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714784.3 | XP_002714830.1 | 7 - 8 | 2218 | 63 | |
| XM_008263437.2 | XP_008261659.1 | 7 - 8 | 2219 | 63 | ||
| XM_008263438.2 | XP_008261660.1 | 7 - 8 | 2220 | 63 | ||
| XM_008263439.2 | XP_008261661.1 | 6 - 7 | 2097 | 63 | ||
| XM_008263440.2 | XP_008261662.1 | 7 - 8 | 2219 | 63 | ||
| XM_008263441.2 | XP_008261663.1 | 7 - 8 | 2218 | 63 | ||
| XM_008263442.2 | XP_008261664.1 | 7 - 8 | 2218 | 63 | ||
| XM_017345339.1 | XP_017200828.1 | 6 - 7 | 2022 | 63 | ||
| XM_017345340.1 | XP_017200829.1 | 7 - 8 | 2224 | 63 | ||
Set Membership: |
|