Search
Search

PNOC| Gene Symbol: PNOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709448.3 | XP_002709494.2 | 2 - 3 | 252 | 80 | |
| XM_008249155.2 | XP_008247377.1 | 2 - 3 | 207 | 80 | ||
| XM_008249163.2 | XP_008247385.1 | 2 - 3 | 270 | 80 | ||
| XM_008249172.1 | XP_008247394.1 | 2 - 3 | 252 | 80 | ||
Set Membership: |
|