Search
Search

ZFP90| Gene Symbol: ZFP90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711637.3 | XP_002711683.1 | 2 - 3 | 234 | 79 | |
| XM_008257520.2 | XP_008255742.1 | 2 - 3 | 272 | 79 | ||
| XM_008257521.2 | XP_008255743.1 | 2 - 3 | 262 | 79 | ||
| XM_008257522.2 | XP_008255744.1 | 2 - 3 | 280 | 79 | ||
Set Membership: |
|