Search
Search

CRADD| Gene Symbol: CRADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711202.3 | XP_002711248.1 | 1 - 2 | 51 | 103 | |
| XM_008256884.2 | XP_008255106.1 | 1 - 2 | 53 | 103 | ||
| XM_017342020.1 | XP_017197509.1 | 1 - 2 | 66 | 103 | ||
| XM_017342021.1 | XP_017197510.1 | 1 - 2 | 58 | 103 | ||
Set Membership: |
|