Search
Search

MSI1| Gene Symbol: MSI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272304.2 | XP_008270526.1 | 4 - 5 | 449 | 76 | |
| XM_017349570.1 | XP_017205059.1 | 4 - 5 | 449 | 76 | ||
| XM_017349571.1 | XP_017205060.1 | 4 - 5 | 449 | 76 | ||
Set Membership: |
|