Search
Search

SMC1B| Gene Symbol: SMC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251933.2 | XP_008250155.1 | 911 | 96 | ||
| XM_017339556.1 | XP_017195045.1 | 911 | 96 | |||
| XM_017339557.1 | XP_017195046.1 | 911 | 96 | |||
Set Membership: |
|