Search
Search

ZNF287| Gene Symbol: ZNF287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718800.3 | XP_002718846.1 | 5 - 6 | 1324 | 85 | |
| XM_008270689.2 | XP_008268911.1 | 5 - 6 | 1181 | 85 | ||
Set Membership: |
|