Search
Search

PIGN| Gene Symbol: PIGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713621.3 | XP_002713667.2 | 25 - 26 | 2703 | 66 | |
| XM_008261431.2 | XP_008259653.1 | 24 - 25 | 2461 | 66 | ||
| XM_017344218.1 | XP_017199707.1 | 26 - 27 | 2773 | 66 | ||
| XM_017344219.1 | XP_017199708.1 | 24 - 25 | 2610 | 66 | ||
Set Membership: |
|