Search
Search

DZIP3| Gene Symbol: DZIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266983.2 | XP_008265205.1 | 32 | 5449 | 85 | |
| XM_008266984.2 | XP_008265206.1 | 32 | 5316 | 85 | ||
| XM_008266985.2 | XP_008265207.1 | 29 | 4932 | 85 | ||
| XM_008266986.2 | XP_008265208.1 | 31 | 4831 | 85 | ||
| XM_017347116.1 | XP_017202605.1 | 31 | 5380 | 85 | ||
Set Membership: |
|