Search
Search

ANKAR| Gene Symbol: ANKAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342982.1 | XP_017198471.1 | 9 - 10 | 2227 | 69 | |
| XM_017342984.1 | XP_017198473.1 | 9 - 10 | 2231 | 69 | ||
| XM_017342985.1 | XP_017198474.1 | 8 - 9 | 1237 | 69 | ||
| XM_017342986.1 | XP_017198475.1 | 6 - 7 | 892 | 69 | ||
Set Membership: |
|