Search
Search

RASSF2| Gene Symbol: RASSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256272.2 | XP_008254494.1 | 9 - 10 | 883 | 58 | |
| XM_008256275.2 | XP_008254497.1 | 8 - 9 | 748 | 58 | ||
Set Membership: |
|