Search
Search

CFAP70| Gene Symbol: CFAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269962.2 | XP_008268184.2 | 8 - 9 | 1026 | 62 | |
| XM_017348708.1 | XP_017204197.1 | 8 - 9 | 1026 | 62 | ||
| XM_017348709.1 | XP_017204198.1 | 8 - 9 | 1027 | 62 | ||
Set Membership: |
|