Search
Search

LOC103352510| Gene Symbol: LOC103352510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275370.2 | XP_008273592.1 | 447 | 102 | ||
| XM_008275371.2 | XP_008273593.1 | 447 | 102 | |||
| XM_017337576.1 | XP_017193065.1 | 339 | 102 | |||
| XM_017337578.1 | XP_017193067.1 | 447 | 102 | |||
Set Membership: |
|