Search
Search

AGBL3| Gene Symbol: AGBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258132.2 | XP_008256354.1 | 3 - 4 | 2527 | 76 | |
| XM_008258135.2 | XP_008256357.1 | 4 - 5 | 2102 | 76 | ||
| XM_008258136.2 | XP_008256358.1 | 4 - 5 | 1499 | 76 | ||
| XM_008258138.2 | XP_008256360.1 | 3 - 4 | 2000 | 76 | ||
| XM_008258140.2 | XP_008256362.1 | 3 - 4 | 2525 | 76 | ||
| XM_017342687.1 | XP_017198176.1 | 3 - 4 | 2525 | 76 | ||
Set Membership: |
|