Search
Search

C4H20orf194| Gene Symbol: C4H20orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710883.3 | XP_002710929.1 | 31 - 32 | 2958 | 107 | |
| XM_008256246.2 | XP_008254468.1 | 31 - 32 | 2955 | 107 | ||
| XM_017341739.1 | XP_017197228.1 | 31 - 32 | 2852 | 107 | ||
Set Membership: |
|