Search
Search

FAM227B| Gene Symbol: FAM227B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269036.2 | XP_008267258.1 | 12 - 13 | 1258 | 74 | |
| XM_008269037.2 | XP_008267259.1 | 11 - 12 | 1156 | 74 | ||
Set Membership: |
|