Search
Search

GPATCH2| Gene Symbol: GPATCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717402.3 | XP_002717448.1 | 4 - 5 | 1122 | 65 | |
| XM_017347781.1 | XP_017203270.1 | 4 - 5 | 1120 | 65 | ||
Set Membership: |
|