Search
Search

PRKAG1| Gene Symbol: PRKAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711089.3 | XP_002711135.1 | 6 - 7 | 444 | 98 | |
| XM_008256409.2 | XP_008254631.1 | 5 - 6 | 334 | 98 | ||
Set Membership: |
|