Search
Search

SLF2| Gene Symbol: SLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718659.3 | XP_002718705.1 | 19 - 20 | 3611 | 87 | |
| XM_008270364.2 | XP_008268586.1 | 19 - 20 | 3850 | 87 | ||
| XM_008270365.2 | XP_008268587.1 | 19 - 20 | 3630 | 87 | ||
Set Membership: |
|