Search
Search

PRUNE1| Gene Symbol: PRUNE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715572.3 | XP_002715618.1 | 7 - 8 | 1184 | 69 | |
| XM_017345902.1 | XP_017201391.1 | 3 - 4 | 334 | 69 | ||
Set Membership: |
|