Search
Search

ERO1A| Gene Symbol: ERO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348298.1 | XP_017203787.1 | 12 - 13 | 1098 | 107 | |
| XM_017348299.1 | XP_017203788.1 | 12 - 13 | 1083 | 107 | ||
| XM_017348300.1 | XP_017203789.1 | 11 - 12 | 1071 | 107 | ||
| XM_017348301.1 | XP_017203790.1 | 12 - 13 | 1098 | 107 | ||
Set Membership: |
|