Search
Search

RBM45| Gene Symbol: RBM45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712309.3 | XP_002712355.1 | 8 - 9 | 1357 | 77 | |
| XM_008258789.2 | XP_008257011.1 | 7 - 8 | 1234 | 77 | ||
| XM_017342954.1 | XP_017198443.1 | 8 - 9 | 1364 | 77 | ||
| XM_017342955.1 | XP_017198444.1 | 8 - 9 | 1361 | 77 | ||
Set Membership: |
|