Search
Search

UROC1| Gene Symbol: UROC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713076.3 | XP_002713122.1 | 5 - 6 | 604 | 54 | |
| XM_017343703.1 | XP_017199192.1 | 5 - 6 | 494 | 54 | ||
| XM_017343704.1 | XP_017199193.1 | 5 - 6 | 531 | 54 | ||
Set Membership: |
|