Search
Search

RPP14| Gene Symbol: RPP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713404.3 | XP_002713450.2 | 3 - 4 | 374 | 93 | |
| XM_008260841.2 | XP_008259063.1 | 3 - 4 | 317 | 93 | ||
| XM_008260842.2 | XP_008259064.1 | 3 - 4 | 380 | 93 | ||
Set Membership: |
|