Search
Search

UCHL5| Gene Symbol: UCHL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717691.3 | XP_002717737.1 | 5 - 6 | 537 | 103 | |
| XM_008268729.2 | XP_008266951.1 | 5 - 6 | 537 | 103 | ||
| XM_008268730.2 | XP_008266952.1 | 5 - 6 | 537 | 103 | ||
Set Membership: |
|