Search
Search

RIT1| Gene Symbol: RIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264332.2 | XP_008262554.1 | 5 - 6 | 562 | 81 | |
| XM_008264333.2 | XP_008262555.1 | 5 - 6 | 575 | 81 | ||
| XM_008264334.2 | XP_008262556.1 | 5 - 6 | 557 | 81 | ||
| XM_008264335.2 | XP_008262557.1 | 5 - 6 | 559 | 81 | ||
Set Membership: |
|