Search
Search

RASGEF1B| Gene Symbol: RASGEF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717085.3 | XP_002717131.1 | 4 - 5 | 591 | 61 | |
| XM_017347474.1 | XP_017202963.1 | 7 - 8 | 689 | 61 | ||
Set Membership: |
|