Search
Search

TRIM45| Gene Symbol: TRIM45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264595.2 | XP_008262817.1 | 4 - 5 | 2720 | 76 | |
| XM_008264596.2 | XP_008262818.1 | 4 - 5 | 2720 | 76 | ||
Set Membership: |
|