Search
Search

BTBD7| Gene Symbol: BTBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349395.1 | XP_017204884.1 | 6 - 7 | 1882 | 62 | |
| XM_017349396.1 | XP_017204885.1 | 6 - 7 | 1883 | 62 | ||
Set Membership: |
|