Search
Search

LRRC19| Gene Symbol: LRRC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708045.3 | XP_002708091.1 | 3 - 4 | 705 | 83 | |
| XM_008251135.2 | XP_008249357.1 | 4 - 5 | 799 | 83 | ||
| XM_008251165.2 | XP_008249387.1 | 4 - 5 | 979 | 83 | ||
Set Membership: |
|