Search
Search

IGF2BP2| Gene Symbol: IGF2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716463.3 | XP_002716509.1 | 1 - 2 | 275 | 63 | |
| XM_002716464.3 | XP_002716510.1 | 1 - 2 | 275 | 63 | ||
Set Membership: |
|