Search
Search

RIPK1| Gene Symbol: RIPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350507.1 | XP_017205996.1 | 534 | 70 | ||
| XM_017350508.1 | XP_017205997.1 | 533 | 70 | |||
| XM_017350509.1 | XP_017205998.1 | 290 | 70 | |||
Set Membership: |
|