Search
Search
| Gene Symbol: ZFX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719982.3 | XP_002720028.1 | 3 - 4 | 750 | 94 | |
| XM_008272461.2 | XP_008270683.1 | 3 - 4 | 801 | 94 | ||
| XM_008272463.2 | XP_008270685.1 | 3 - 4 | 714 | 94 | ||
| XM_017349825.1 | XP_017205314.1 | 3 - 4 | 758 | 94 | ||
| XM_017349826.1 | XP_017205315.1 | 2 - 3 | 630 | 94 | ||
| XM_017349827.1 | XP_017205316.1 | 2 - 3 | 630 | 94 | ||
Set Membership: |
|