Search
Search

TAF2| Gene Symbol: TAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710750.3 | XP_002710796.1 | 22 - 23 | 3142 | 68 | |
| XM_008255856.2 | XP_008254078.1 | 22 - 23 | 3142 | 68 | ||
| XM_008255857.2 | XP_008254079.1 | 22 - 23 | 3142 | 68 | ||
| XM_008255858.2 | XP_008254080.1 | 22 - 23 | 3142 | 68 | ||
| XM_008255859.2 | XP_008254081.1 | 18 - 19 | 2397 | 68 | ||
| XM_017341500.1 | XP_017196989.1 | 22 - 23 | 3142 | 68 | ||
Set Membership: |
|