Search
Search

GIN1| Gene Symbol: GIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713944.3 | XP_002713990.1 | 7 - 8 | 1494 | 108 | |
| XM_008261985.2 | XP_008260207.1 | 7 - 8 | 1635 | 108 | ||
| XM_008261986.2 | XP_008260208.2 | 7 - 8 | 1976 | 108 | ||
Set Membership: |
|